Would you like to log in again?
Research® plus Multi Channel Mechanical Pipettors, Eppendorf
Supplier: Eppendorf
Research® plus
3125000087 3125000095 3125000109 3125000079 3125000222
76311-128EA 1938.2 USD InStock
76311-128 76311-126 76311-124 MSPP-312500022 76311-122
Research® plus Multi Channel Mechanical Pipettors, Eppendorf
Pipettes Micropipettes
Versatile, ergonomic pipettors with innovative design, meeting the highest needs in precision and accuracy.
  • Extremely lightweight
  • Innovative re-designed principle of piston-stroke pipetting, piston made of organic polymer Fortron® with great chemical, thermal and mechanical resistance
  • Lowest aspirating, dispensing, blow-out and tip ejection forces
  • Spring loaded retractable nose cone to ensure tight seal and prolong nose cone life, as well as exact tip alignment
  • Individual channel removal for quick and easy servicing
  • Fully autoclavable
Designed and constructed for general laboratory and research applications in Life Science. Typical applications include: Food & beverages, forensic, cosmetics, agriculture, animal studies, chemical analytics, research applications at medical and academic facilities.
Certifications: Without CE marking.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty