Would you like to log in again?
LoBind® Protein Microcentrifuge Tubes, Eppendorf
Supplier: Eppendorf
Protein LoBind
0030108450
MSPP-030108450EA 152 USD InStock
MSPP-030108450
LoBind® Protein Microcentrifuge Tubes, Eppendorf
Tubes Microcentrifuge Tubes
Maximize protein recovery with Protein LoBind® Tubes.
  • Prevent sample loss due to sample-surface binding
  • Unique, two-component polymer mix that helps to create a hydrophilic surface
  • Protein recovery rate and downstream analysis is significantly improved
With standard vessels, over 90% of the sample can be lost within 24 hours due to binding to the plastic surface. With Eppendorf LoBind Tubes there is no need to worry about losing precious samples any longer.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty