Would you like to log in again?
ep Dualfilter T.I.P.S.® Filter Tips (GLP), Eppendorf
Supplier: Eppendorf
epT.I.P.S® Dualfilter
0030078586 0030078586
76423-882PK 124.16 USD InStock
CA76423-882 76423-882
ep Dualfilter T.I.P.S.® Filter Tips (GLP), Eppendorf
Pipette Tips
The unique double layer (two-phase) filter for ultimate protection of pipette and sample against unintentional and unnoticed contamination by aerosols, drops and splashes.
  • Two filter layers provide dual protection against aerosols and biomolecules to guard your pipette and sample
  • Free of PCR inhibiting additives provide for reliable downstream results
  • Available in PCR clean/sterile (sterile and pyrogen-free)
  • Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality
  • Modern slim rack design with reclosable lid
With defined flow dynamics, low wettability, and high thermal stability they are certified pyrogen-free and PCR clean.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty