Would you like to log in again?
SnapTec® 50 Conical Tubes, Eppendorf
Supplier: Eppendorf
SnapTec®
0030118669 0030118650 0030118677
76534-800EA 124.28 USD InStock
76534-800 76534-798 76534-796
SnapTec® 50 Conical Tubes, Eppendorf
Tubes Centrifuge Tubes
Eppendorf conical tubes SnapTec® 50 enable safe opening and closing, minimise contamination risk and are implementable in existing lab equipment. Allowing very tight closure, sample loss is minimised making them the ideal solution for cell culture and cell biology applications, and sample preparation in microbiology and molecular biology.
  • Premium USP class - 6 raw material and no use of slip agents, plasticisers, and biocides during manufacturing ensure highest sample integrity, high g-Safe® centrifugation stability, and optimal sample and pellet visibility
  • Cap with flattened and grooved sides supports a user-friendly ergonomic handling, enables a stable upright positioning, and minimises contamination risk
  • Fast opening and closing in routine multistep protocols
  • Autoclavable at 121 °C for 20 minutes
  • Reliable and safe labelling due to a large writing area on the tube and due to a flat and light-coloured cap
  • High g-Safe centrifugation stability up to 17000×g
  • Precise dimensions enable maximum compatibility with centrifuge rotors, mixers and shakers
The Conical Tubes SnapTec® 50 extend the range of the Eppendorf Tubes®. Safe design and easy handling ensure best sample processing. They represent an optimised solution for cell culture and cell biology applications as well as for sample preparation protocols in microbiology and molecular biology laboratories. Whether centrifugation, mixing, or other applications, the Eppendorf Conical Tubes 15 and 50 ml - available either with SnapTec® cap and screw cap or as smart versions like LoBind® or amber - will complete your workflows in an excellent way.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty