Would you like to log in again?
J.T.Baker® BAKERBOND® Q2100 New Generation UHPLC and HPLC Columns, Avantor®
Supplier: Avantor
J.T.Baker® BAKERBOND®
7378-24
76470-960EA 884.96 USD InStock
76470-960
J.T.Baker® BAKERBOND® Q2100 New Generation UHPLC and HPLC Columns, Avantor®
Chromatography Columns
BAKERBOND® Q2100 LC phases have been specially designed to solve the challenges of most routine samples. They can also be a good starting point for new method development projects. With options of four widely used chemistries and three particle sizes BAKERBOND® Q2100 can be a useful addition to your analytical toolkit.
  • High efficiency UHPLC/HPLC columns
  • Based on high purity spherical silica
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty