Would you like to log in again?
Yeast Mating Factor alpha (from Yeast)
Supplier: GenScript Biotech
RP01002
76510-680EA 125.4 USD InStock
76510-680
Yeast Mating Factor alpha (from Yeast)
Proteins and Peptides
This alpha factor pheromone arrests yeast in the G1 phase of their cell cycle. It helps to induces the expression of mating gene, changes in nuclear architecture and consequently polarizes the growth toward the mating partner.
STE2 encodes the presence of alpha factor pheromone receptor on mating type A cells in yeast.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty