Would you like to log in again?
Hotplate Stirrers, GHS-550 Group, Globe Scientific
Supplier: Globe Scientific
GHS-550HSP GHS-550HS GHS-550S
76595-392EA 733.33 USD InStock
76595-392 76595-394 76595-396
Hotplate Stirrers, GHS-550 Group, Globe Scientific
Stirrers Magnetic Hotplate Stirrers
The GHS-550 group of Hotplates, Magnetic Stirrers and Magnetic Stirrer Hotplates offer safety and precision for laboratory heating and mixing applications. The compact and low-profile design saves valuable benchtop space and a 7" chemical resistant glass ceramic working surface ensure years of service and easy cleanup.
  • Simple, intuitive operation
  • Maintenance-free brushless DC motor
  • Small footprint saves valuable bench space
  • Powerful 1000 W heater
  • Easy-to-clean glass ceramic surface
  • Easy-to-read LCD or LED display for simple, clear control
  • Powerful magnetic coupling for stirring up to 20 L
  • Optional external temperature sensor control
Setting the heat and stirring speed is extremely intuitive with an LED display and control knobs, inputting to a microprocessor-controlled circuit for accurate, stable control of heating and stirring.
The GHS-550HSP model replaces the LED with an LCD display for precise monitoring of speed and temperature. This hot plate magnetic stirrer also includes a built-in PID temperature control heating with overheating protection. A RS232 port offers remote function for communication, PC control and data transmission (software not included).
The Magnetic Stirrer and Hotplate Magnetic Stirrer are designed to provide a strong magnetic coupling for powerful mixing that is enabled by the maintenance free brushless DC motor. The stirrers can mix up to 10 L of water (GHS-550HS, GHS-550S) or 20 L of water (GHS-550HSP), which allows for excellent homogeneity of your solution.
The Hotplate and Hotplate Magnetic Stirrer models are designed with a powerful 1000 W heater that provides temperatures up to 550 °C. To gain even more accurate temperature control within 0.2 °C, an optional PT1000 sensor and retort stand are available to measure directly in the vessel. A 'hot' warning will flash if the work plate temperature is above 50 °C. Over temperature protection is included should temperatures reach 580 °C.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty