Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

G418 disulfate

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning
Synonyms: G-418 disulfateG-418 disulphateG418 disulfateG-418 sulphateG418 sulphateG-418 sulfateGeneticin®

0215878294
76461-468EA 578.69 USD InStock
76461-468
G418 disulfate
G418 disulphate


Formula: C₂₀H₄₄N₄O₁₈S₂
MW: 692.72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058314
CAS Number: 108321-42-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty