Would you like to log in again?
HyClone™ CHO Cell Culture Platform Media, SFM4CHO Utility Powder (HyClone products (Cytiva))
Supplier: HyClone products (Cytiva)
HyClone™
SH30517.01 SH30517.01
76635-850EA 250.78 USD InStock
76635-850 CA76635-850
HyClone™ CHO Cell Culture Platform Media, SFM4CHO Utility Powder (HyClone products (Cytiva))
Cell Culture Media
HyClone™ SFM4CHO-Utility media is ideally suited for the cost-effective manufacturing of recombinant proteins for academic and industrial research, genomics and proteomics, drug target screening and validation, and manufacturing of preclinical lots. It is formulated using a proprietary lipid complexing process for enhanced stability and broad-spectrum growth promotion.
  • Supports the DHFR selection/amplification system
  • Defined protein-free formulation
  • Metabolically designed for high cell yield and recombinant protein production
  • Supports multiple CHO clones
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured under cGMP
SFM4CHO-Utility does not contain phenol red. It has been designed to support the DHFR selection/amplification system. This protein-free media has been successfully tested in a variety of culture systems including T-flasks, spinner and shaker flasks, roller bottles, and bioreactors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty