Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethanol 200 Proof USP, pure

Supplier: TILLEY CHEMICAL CO., INC
Danger
Synonyms: Ethyl alcohol

DNA029 DNA029TP DNA070 DNA070TP
TLDNA029TPEA 1010186.31 USD InStock
TLDNA029TP TLDNA070 TLDNA029 TLDNA070TP
Ethanol 200 Proof USP, pure
Ethanol


Formula: C₂H₅OH
MW: 46.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty