Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water nuclease-free

Supplier: OMEGA BIO-TEK
Synonyms: Dihydrogen oxide

PD092
76539-260EA 25.19 USD InStock
76539-260
Water nuclease-free
Water
Caution: For Research Use Only. Not for use in diagnostic procedures.


Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 12,10175
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty