Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

E-Z 384 Plant DNA HT Kits, Omega Bio-tek®

Supplier: Omega Bio-Tek
E-Z 384
D1341-00 D1341-01
76539-230EA 785.62 USD InStock
76539-230 76539-228
E-Z 384 Plant DNA HT Kits, Omega Bio-tek®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The E-Z 384 Plant DNA HT kit enables rapid and high–throughput isolation of genomic DNA from a wide variety of plant tissue samples. Up to 768 fresh, frozen, or dried plant tissue samples rich in polyphenols, polysaccharides or having a lower DNA content can be processed in approximately 1.5 hours when processing two plates in parallel.

  • Productivity – Allows processing of up to 768 samples in parallel
  • Robust – Reliable results from a variety of sample types
  • Safe – No organic extractions
  • Performance – High-quality DNA sufficient for most applications

The system combines the reversible nucleic acid-binding properties of the HiBind® matrix with the speed and versatility of silica plate technology to eliminate polysaccharides, phenolic compounds, and enzyme inhibitors from plant tissue lysates. The purification procedure relies on the well-established properties of the cationic detergent, cetyltrimethyl ammonium bromide (CTAB), in conjunction with a unique binding system to provide high-quality DNA. The system eliminates the need for chloroform extractions traditionally associated with CTAB-based lysis methods. The eluted DNA is inhibitor-free and ready for downstream applications such as genotyping, plant breeding, population level analyses, Next Generation Sequencing, PCR, and more.

Specifications
Starting Material: Fresh, frozen, or dried plant tissue samples rich in polysaccharides, polyphenols, or those having a low DNA content
Starting Amount: 30 mg fresh/10 mg dry plant sample per well
Elution Volume: 65 µl
Processing Mode: Manual (Centrifugation or Vacuum)
Throughput: 1 - 768 (process two 384-plates in parallel)
Nucleic Acid Binding Technology: 384-well silica filter plate
Processing time: 1.5 h
Downstream Application(s): Genotyping, plant breeding, population level analyses, next generation sequencing, PCR, and more.

Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty