Would you like to log in again?
SCI550-S LED Digital Hotplate Stirrer, Scilogex
Supplier: Scilogex
813221009999
MSPP-813221009EA 710.32 USD InStock
MSPP-813221009
SCI550-S LED Digital Hotplate Stirrer, Scilogex
Stirrers Magnetic Hotplate Stirrers
This SCI550-S provides powerful heating and stirring with LED digital display of the set temperature and real-time temperature display. LED heating and analog stirring are simply controlled turning the respective control knobs.
  • LED display for precise monitoring of temperature
  • A wide speed range of 50 to 1500rpm
  • Heat only, stir only or heat and stir together functions
  • Max. Heating plate temperature 550 °C
  • Low temperature stability
  • High magnetic adhesion
Electronic speed control for constant speed even during changes in load. External temperature control is possible by connecting the temperature sensor (PT1000) with accuracy at ±0.2 °C.
'HOT' warning will flash if the work plate temperature is above 50 °C even when the hotplate is turned off.
cTUVus approved for UL and CSA. 2 year warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty