Would you like to log in again?
Twin.tec® PCR Plates - PCR Plates, Eppendorf
Supplier: Eppendorf
Twin.tec®
0030133331 0030129350 0030129326 0030129300 0030133390 0030129318 0030133366 0030129342 0030129334
89136-706PK 137.37 USD InStock
89136-706 89136-704 89136-694 89136-708 89136-686 89136-696 89136-712 89136-702 89136-710
Twin.tec® PCR Plates - PCR Plates, Eppendorf
Microplates PCR Plates
The Eppendorf twin.tec® PCR plate combines extremely thin-walled polypropylene wells for optimal heat transfer to the sample, with robust polycarbonate frames for high rigidity. With raised well rims, twin.tec plates enable highly effective sealing to reduce the risk of cross-contamination.
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid and torque-resistant polycarbonate frame
  • Improved well-to-well tolerance
  • Cut-away corner and alphanumeric grid-referencing
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNase, RNase and PCR inhibitors (PCR clean)
  • Available with barcode (upon request), for skirted and semi-skirted only
Twin.tec plates are available in many different formats to suit different workflows and applications:
96-well and 384-well formats
Skirted, semi-skirted, and unskirted
Divisible plates to avoid waste in small assays
Individually wrapped forensic DNA grade plates
Microbiology plates
Barcoded plates
All Eppendorf twin.tec PCR plates are ‘PCR clean’, having been batch-tested and certified by an independent laboratory to be free of a range of common PCR contaminants, including DNA, DNAse, RNAse, and PCR inhibitors.
These plates are used in various applications standard PCR, real-time PCR, high throughput PCR, small volume sample handling.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty