Would you like to log in again?
TI-Innovator Water Pump Pack
Supplier: D&H
TI-Innovator
WATERPUMP5PK
470348-852EA 58.99 USD InStock
470348-852
TI-Innovator Water Pump Pack
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Control a water pump with your TI-Innovator hub system.
  • Practice programming skills by physically controlling a small water pump
  • Students see the physical results of their code
This submergible water pump is controlled by your TI-Innovator hub, providing a unique and interesting opportunity to use programming to develop useful, real-world applications. Build and program your Internet of Things (IoT) in your classroom using the TI-Innovator hub system and your existing TI-Nspire or TI-84 Plus CE technology.
Delivery information: Pack includes five water pumps and five sets of tubing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty