Would you like to log in again?
TI-Innovator External Battery
Supplier: D&H
TI-Innovator
TIEXTERNALBTRYK STEMBT/AC/2L1
470348-844EA 19.99 USD InStock
CA-470350-666 470348-844
TI-Innovator External Battery
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Provide power for output components for your TI-Innovator Hub system.
  • Enable devices that need more power than the TI-Innovator Hub
Some physical computing input and output components require relatively high current. Expand your IoT and programming capabilities with this recommended accessory for the TI-Innovation Hub. The External Battery provides a source of power when using components that require extra power.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty