Would you like to log in again?
TI-Innovator MOSFET Module
Supplier: D&H
TI-Innovator
MOSFETMOD5PK
470348-856EA 32.99 USD InStock
470348-856
TI-Innovator MOSFET Module
Educational Materials Physics Educational Materials Applied Physics Learning Activities
MOSFET transistor for your TI-Innovator system.
  • Design devices and advanced experiments with power control or signal amplification
MOSFET transistors are power control components - they can be used to amplify signals. The TI-Innovator MOSFET module comes mounted on a printed circuit board for easy handling, and includes a AA battery holder. Build and program your Internet of Things (IoT) in your classroom using the TI-Innovator hub system and your existing TI-Nspire or TI-84 Plus CE technology.
Delivery information: Pack includes five MOSFET modules and five AA battery holders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty