Would you like to log in again?
TI-Innovator Hall Effect Module
Supplier: D&H
TI-Innovator
TIHALLSENSMOD5P
470348-878EA 42.99 USD InStock
470348-878
TI-Innovator Hall Effect Module
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Hall Effect sensor for your TI-Innovator system.
  • Design devices and advanced experiments with a proximity sensor
The Hall Effect is a voltage caused in a conductor by a perpendicular currents and magnetic fields, allowing the sensor to measure proximity and motion in many types of systems. The TI-Innovator Hall Effect Module comes mounted on a printed circuit board for easy handling, with a cable to connect it to the TI-Innovator Hub for programming. Build and program your Internet of Things (IoT) in your classroom using the TI-Innovator Hub system and your existing TI-Nspire or TI-84 Plus CE technology.
Delivery information: Pack includes five hall sensors and cables.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty