Would you like to log in again?
TI-Innovator Light Sensor
Supplier: D&H
TI-Innovator
LIGHTSENSMOD5PK
470348-848EA 20.99 USD InStock
470348-848
TI-Innovator Light Sensor
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Light sensor for your TI-Innovator hub system.
  • Collect light intensity data as inputs for programming
  • Real-world applications for your TI-Innovator hub
The TI-Innovator light sensor detects the light intensity of the environment. It comes mounted on a printed circuit board for easy handling, with a cable to connect it to the TI-Innovator Hub for programming. Build and program your Internet of Things (IoT) in your classroom using the TI-Innovator Hub system and your existing TI-Nspire or TI-84 Plus CE technology.
Delivery information: Pack includes five light sensors and cables.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty