Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dodecyl-β-D-glucopyranoside

Supplier: MilliporeSigma
Synonyms: n-Dodecyl β-D-glucopyranosideDodecyl glucoside

324351-1GM 324351-1GM
80056-362EA 293.05 USD InStock
80056-362 CA80056-362
Dodecyl-β-D-glucopyranoside
Dodecyl β-D-glucopyranoside


Formula: C₁₈H₃₆O₆
MW: 348.48 g/mol
MDL Number: MFCD00063298
CAS Number: 59122-55-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty