Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside

Supplier: MilliporeSigma
Synonyms: 4-Nitrophenyl 2-acetamido-2-deoxy-beta-D-glucopyranoside4-Nitrophenyl N-Acetyl-β-D-glucosaminide

487052-1GM 487052-1GM
80057-138EA 233.73 USD InStock
80057-138 CA80057-138
4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside
4-Nitrophenyl-2-acetamido-2-deoxy-beta-D-glucopyranoside


Formula: C₁₄H₁₈N₂O₈
MW: 342.30 g/mol
MDL Number: MFCD00063696
CAS Number: 3459-18-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty