Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Lauroylsarcosine sodium salt

Supplier: MilliporeSigma
Synonyms: SarkosylSodium lauroyl sarcosineSodium N-lauroylsarcosinate

428010-5GM 428010-5GM
80058-304EA 314.05 USD InStock
CA80058-304 80058-304
N-Lauroylsarcosine sodium salt
N-Lauroylsarcosine sodium salt


Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
MW: 293.38 g/mol
MDL Number: MFCD00042728
CAS Number: 137-16-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty