Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Octyl-β-D-thioglucopyranoside, ULTROL®

Supplier: MilliporeSigma
Synonyms: (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triolN-Octyl-1-thio-Beta-D-glucopyranosideOTG1-S-Octyl-β-D-thioglucopyranosideOctyl-thioglucoside

494461-5GM 494461-5GM
80058-532EA 483.85 USD InStock
CA80058-532 80058-532
Octyl-β-D-thioglucopyranoside, ULTROL®
Octyl β-D-thioglucopyranoside


Formula: C₁₄H₂₈O₅S
MW: 308.44 g/mol
MDL Number: MFCD00012189
CAS Number: 85618-21-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty