Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(-)-Sorbose

Supplier: MilliporeSigma
Synonyms: (3S,4R,5S)-1,3,4,5,6-Pentahydroxyhexan-2-one

567692-1KG 567692-250GM 567692-250GM 567692-1KG
80109-424EA 486.77 USD InStock
80109-424 CA80109-422 80109-422 CA80109-424
L(-)-Sorbose
L(-)-Sorbose


Formula: C₆H₁₂O₆
MW: 180.16 g/mol
MDL Number: MFCD00151097
CAS Number: 87-79-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty