Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Arabinose

Supplier: MilliporeSigma
Synonyms: (2S,3R,4R)-2,3,4,5-Tetrahydroxypentanal

178670-25GM 178670-25GM
80502-266EA 93.82 USD InStock
80502-266 CA80502-266
D-(-)-Arabinose
D(-)-Arabinose


Formula: C₅H₁₀O₅
MW: 150.13 g/mol
MDL Number: MFCD00135608
CAS Number: 10323-20-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty