Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polymyxin B sulphate in aqueous solution cell culture tested, sterile-filtered

Supplier: MilliporeSigma

420413-20ML 420413-20ML
82603-000EA 423.18 USD InStock
82603-000 CA82603-000
Polymyxin B sulphate in aqueous solution cell culture tested, sterile-filtered
Polymyxin B sulphate


Formula: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
MW: 1385.61 g/mol
MDL Number: MFCD00131991
CAS Number: 1405-20-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty