Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tenovin-1

Supplier: MilliporeSigma
Synonyms: N-((4-Acetamidophenyl)carbamothioyl)-4-(tert-butyl)benzamide

580566-10MG 580566-10MG
82603-346EA 96.43 USD InStock
82603-346 CA82603-346
Tenovin-1
Tenovin-1


Formula: C₂₀H₂₃N₃O₂S
MW: 369,48 g/mol
MDL Number: MFCD02914760
CAS Number: 380315-80-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty