Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water, LiChrosolv® tested for UHPLC-MS, Supelco®

Supplier: MilliporeSigma
Synonyms: Dihydrogen oxide

1.03728.2002 1.03728.1002
EM1.03728.1002EA 144 USD InStock
EM1.03728.1002 EM1.03728.2002
Water, LiChrosolv® tested for UHPLC-MS, Supelco®
Water


Formula: H₂O
MW: 18.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty