Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oxalic acid c(CsH2O4) = 0.005 mol; standardised against Potassium permangante solution, Titripur®, Supelco®

Supplier: MilliporeSigma

1.60329.1000
EM1.60329.1000EA 77 USD InStock
EM1.60329.1000
Oxalic acid c(CsH2O4) = 0.005 mol; standardised against Potassium permangante solution, Titripur®, Supelco®
Oxalic acid


Formula: C₂H₂O₄
MW: 90.04 g/mol
MDL Number: MFCD00002573
CAS Number: 144-62-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty