Would you like to log in again?
Life/form® Cathed Trainer Set
Supplier: NASCO HEALTHCARE INC. SE
Life/form®
LF01029
470348-678EA 2805.8 USD InStock
470348-678
Life/form® Cathed Trainer Set
Models Human Biology Models
Easily see the catheterization process.
  • Male and female models
  • Clear surface to visualize progress
  • 5-year warranty
These unique Life/form® simulators combine visual and tactile learning sensations of catheterization in one compact unit. The mid-sagittal dissections allow detailed study of the internal anatomy and provide students an opportunity to follow what happens when they catheterize a patient.
The removable plastic cover identifies key anatomical structures. With practice on Cath-Ed 1 male simulator, normal restrictions at the bulbous urethra and bladder sphincter can be detected and identified. This simulator does not have a functional bladder.
The Cath-Ed 2 provides practice in female catheterization. The vulva has been molded intact to facilitate identifying the urethra and proper spreading of the labia. A functional bladder has been incorporated to allow simulated urine to flow when the catheter is inserted properly. A Foley catheter can be demonstrated and seen through the viewing port.
Delivery information: Set includes: a hard carrying case, lubricant, one catheter, 50cc bulb, syringe, instruction manual, and five-year warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty