Would you like to log in again?
Simulaids® Pediatric ALS Trainer
Supplier: NASCO HEALTHCARE INC. SE
Simulaids®
101-090
470335-340EA 1548.49 USD InStock
470335-340
Simulaids® Pediatric ALS Trainer
Models Human Biology Models
Pediatric life support.
  • Soft carry bag included
  • Latex-free
Features include bag valve mask ventilation, intubation head, oral or nasal intubation, Sellick's Maneuver, accepts NG tube, LMA airways, four lead monitoring, brachial pulse, external chest compressions, jaw thrust, IV sites in hand and arm, two I/O legs for intraosseous needle insertion and aspiration of bone marrow, scalp vein for palpation, cranial sutures, and fontanels.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty