Would you like to log in again?
Drug Testing Cup, Clarity Diagnostics
Supplier: Clarity Diagnostics
CD-CDOA-4104
76329-455CS 2021.09 USD InStock
76329-455
Drug Testing Cup, Clarity Diagnostics
Drug Tests
Clarity CLIA Waived Drug Test Cup 10 Panel. Tests for the following: AMP, BAR, BZO, COC, MAMP (MET), MTD, OPI2000 (OPI), OXY, PCP, THC
  • Built-In Temperature Strip
  • Results in 5 Minutes
  • Screw Top Lid for a No-Leak Cup
  • 510K Cleared and CLIA Waived
They reduce retesting and repeats by identifying suspected adulteration in one step process.
Certifications: CLIA Waived
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty