Would you like to log in again?
HEX-dT Phosphoramidite, AAT Bioquest
Supplier: AAT Bioquest
6210 6211
76644-684EA 194.16 USD InStock
76644-684 76644-686
HEX-dT Phosphoramidite, AAT Bioquest
Protein Linking and Labeling Systems
HEX-dT phosphoramidite provides a valuable tool to add the HEX tag to a desired location of an oligo sequence.
  • DMT group allows quantification
HEX-dT phosphoramidite contains a DMT group to allow quantification of coupling. It can be inserted into a target sequence as a replacement for a dT residue.
It is complimentary to the commonly used 6-HEX phosphoramidite that contains no DMT group and can only be added at the 5'-terminus, thus terminating synthesis.
Packaging: Plastic vial.
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty