Would you like to log in again?
Welch Allyn® ELI 280 Resting ECG
Supplier: Healthcare Products
Welch Allyn®
WELELI280-CAA-AAFAD WELELI280-AAA-AAFBD
470347-196EA 14088.33 USD InStock
470347-196 470347-170
Welch Allyn® ELI 280 Resting ECG
Data Loggers
Multichannel visualization.
  • 10.1" color touchscreen presents all 12 leads of data simultaneously
  • Widescreen display improves navigation, data entry and electrode verification
  • Supports pacemaker spike detection
  • Includes hook-up confirmation by detection and display of lead reversals
The ELI 280 Electrocardiograph provides comprehensive functionality in a portable, touchscreen device. The easy to use device is designed to meet the demands of high-volume ECG environments in both acute care and physician office environments. The widescreen data layout allows for easy menu navigation, fast entry of patient information and clear verification of proper electrode connections. It offers proven ECG performance, digital integration and VERITAS algorithm accuracy with more than 30 years of expertise for a more confident interpretation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty