Would you like to log in again?
DY547 CEP (DyLight 547 Phosphoramidite), AAT Bioquest
Supplier: AAT Bioquest
DyLight
6074
76644-668EA 483.14 USD InStock
76644-668
DY547 CEP (DyLight 547 Phosphoramidite), AAT Bioquest
Protein Linking and Labeling Systems
DY547 CEP (DyLight 547 phosphoramidite) is an excellent replacement to the commonly used Cy3 phosphoramidite.
  • High stability
  • Oligo deprotection can be carried out at room temperature
DY547 CEP appears to be more stable than Cy3 phosphoramidite. As Cy3 and Cy5 phosphoramidites, the deprotection of oligos containing DY547 CEP may be carried out with ammonium hydroxide at room temperature, regardless of the base protecting groups on the monomers used.
It is always prudent to use monomers with base labile protecting groups to limit the exposure time to 2 hours or less at 65 °C during deprotection.
Packaging: Plastic vial.
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty