Would you like to log in again?
DY677 CEP (DyLight 677 Phosphoramidite), AAT Bioquest
Supplier: AAT Bioquest
DyLight
6078
76644-672EA 677.24 USD InStock
76644-672
DY677 CEP (DyLight 677 Phosphoramidite), AAT Bioquest
Protein Linking and Labeling Systems
DY677 CEP (DyLight 677 phosphoramidite) is an excellent replacement to the commonly used Cy5.5 phosphoramidite.
  • High stability
  • Oligo deprotection can be carried out at room temperature
DY677 CEP appears to be more stable than Cy5.5 phosphoramidite. As Cy5.5 phosphoramidite, the deprotection of oligos containing DY677 CEP may be carried out with ammonium hydroxide at room temperature, regardless of the base protecting groups on the monomers used.
It is always prudent to use monomers with base labile protecting groups to limit the exposure time to 2 hours or less at 65 °C during deprotection.
Packaging: Plastic vial.
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty