Would you like to log in again?
VivaGuard® Non-Contact Infrared Thermometer, Germaine Laboratories
Supplier: Germaine Laboratories
VivaGuard®
HA-650
76670-426EA 37.65 USD InStock
76670-426
VivaGuard® Non-Contact Infrared Thermometer, Germaine Laboratories
Thermometers Digital Thermometers
Reliable, fast results in one second.
  • Easy to use!
  • Advanced infrared technology provides quick and accurate measurements in one second
  • Features wide body temperature measuring range between 93.2...109.4 °F
  • Offers reliable results in a wide environmental temperature range at 50.0...104.0 °F
  • Non-contact = safer measurements
  • No measuring sensor cover is needed
  • Minimizes the risk of spread of disease and cross-contamination
  • Eliminates the need for cleaning between each person
  • Features red high fever alert above 101.3°
  • User-friendly design offers comfortable grip
  • Switchable between °F/°C and beep/silent options
  • 2 AAA batteries included
Hold the non-contact thermometer 1 to 5 cm (0.4 to 2") away from the center of forehead to obtain measurement. Five-year shelf life.
Delivery information: One shipping box. Fast shipping.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty