Would you like to log in again?
Flux® Heavy-Duty Drum Pump Motors, TEFC
Supplier: Flux Pumps
F458-1 230V F458-1 120V
MFLX07076-20EA 1778.2 USD InStock
MFLX07076-20 MFLX07076-25
Flux® Heavy-Duty Drum Pump Motors, TEFC
Pumps Drum/Transfer Pumps
Configurable to your requirements.
  • High-viscosity tube set handles fluids with viscosities to 50,000 cp
Heavy-duty motors are best for applications requiring high output head. Motors have a built-in overload switch with manual rest to avoid the danger of automatic restarting.
Order tube set and accessories separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty