Would you like to log in again?
Vacuum Adapters, Promega®
Supplier: Promega
A1331
PAA1331EA 14.85 USD InStock
PAA1331
Vacuum Adapters, Promega®
Adapters Adapters, Joints with Tube Connectors
Vacuum adapters are required for use with the Wizard® SV minicolumns for vacuum-based protocols.
  • Enables use of vacuum-based plasmid miniprep protocol
Component of the Wizard® Plus SV Minipreps DNA Purification Systems A1340 and A1470.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty