Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid fuming 37% in water, pure

Supplier: Thermo Scientific Chemicals
Danger

12462-0010
TS12462-0010EA 51 USD InStock
TS12462-0010
Hydrochloric acid fuming 37% in water, pure
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –35 °C
Density: 1.18 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear solution
Color scale ≤10 APHA
Titration with NaOH 35.0 to 39.0 %
Sulfate (SO4) ≤100 ppm
Free chlorine ≤10 ppm (Cl)
Iron (Fe) ≤5 ppm
Lead (Pb) ≤2 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty