Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetramethylsilane 99%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: TMS

42733-1000
TS42733-1000EA 100 USD InStock
TS42733-1000
Tetramethylsilane 99%, AcroSeal™
Tetramethylsilane


Formula: C₄H₁₂Si
MW: 88.22 g/mol
Boiling Pt: 26.6 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0.64 g/cm³
Flash Pt: –27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008274
CAS Number: 75-76-3
UN: 2749
ADR: 3,I
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3575 to 1.3595 (20 °C, 589 nm)
GC ≥98.5 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty