Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyethylene glycol 4,000 ∼ Mn 4000

Supplier: Ambeed
Warning
Synonyms: PEG 4,000PEG 4000

A139464-25G A139464-500G A139464-100G A139464-1000G
76647-010EA 5.15 USD InStock
76647-010 76647-012 76647-008 76647-006
Polyethylene glycol 4,000 ∼ Mn 4000
Polyethylene glycol 4,000
Caution: for Research Use Only


Formula: H(OCH₂CH₂)nOH
MW: 4000 g/mol
MDL Number: MFCD01779612
CAS Number: 25322-68-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty