Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5,5'-(Hexafluoropropane-2,2-diyl)bis(2-(allyloxy)aniline) 97%

Supplier: Ambeed
Warning

A1349604-10G
76658-106EA 94.76 USD InStock
76658-106
5,5'-(Hexafluoropropane-2,2-diyl)bis(2-(allyloxy)aniline) 97%
5,5'-(Hexafluoropropane-2,2-diyl)bis(2-(allyloxy)aniline)
Caution: for Research Use Only


Formula: C₂₁H₂₀F₆N₂O₂
MW: 446.3861 g/mol
MDL Number: MFCD32708902
CAS Number: 2573217-07-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty