Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-4-(tert-butyl)-2-iodobenzene 95%

Supplier: Ambeed
Warning
Synonyms: 1-Bromo-4-(1,1-dimethylethyl)-2-iodo-benzene

A757255-5G
76652-616EA 276.04 USD InStock
76652-616
1-Bromo-4-(tert-butyl)-2-iodobenzene 95%
1-Bromo-4-(tert-butyl)-2-iodobenzene
Caution: for Research Use Only


Formula: C₁₀H₁₂BrI
MW: 339.0108 g/mol
MDL Number: MFCD28400709
CAS Number: 916747-51-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty