Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium stearate 98%

Supplier: Ambeed
Warning
Synonyms: Stearic acid lithium saltLithium Stearate

A920970-100G A920970-1000G A920970-25G A920970-500G
76655-390EA 52.53 USD InStock
76655-390 76655-392 76655-394 76655-396
Lithium stearate 98%
Lithium stearate
Caution: for Research Use Only


Formula: C₁₈H₃₅LiO₂
MW: 290.42 g/mol
MDL Number: MFCD00042032
CAS Number: 4485-12-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty