Would you like to log in again?
Astrolabe Kit
654-0015
470350-118EA 23.09 USD InStock
470350-118
Astrolabe Kit
Educational Materials Earth Science Educational Materials Astronomy Learning Activities
The easy-to-assemble astrolabe kit can be used to enrich our understanding of astronomy and mathematics.
  • Star calculations
  • Measure altitude of distant objects
  • Discover time by the sun and/or stars
  • Calculate sunrise
  • Determine latitude, and more
This Renissance invention revolutionized how humans performed astronomical calculations. Now you, too, can own one of these marvelous instruments and learn how to use it. Created by Dr. Jim Lattis of the university of Wisconsin-Madison and Dr. Sara Schechner of harvard university, this new astrolabe kit is based on an astrolabe made by Jean Fusoris of Paris around A.D. 1400.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty