Would you like to log in again?
Celestial Sphere Kit
654-0035 654-0030
470350-100PK 98.09 USD InStock
470350-100 470350-132
Celestial Sphere Kit
Educational Materials Earth Science Educational Materials Astronomy Learning Activities
With the celestial sphere, students are able to model the apparent daily motions of the stars and the sun for a variety of latitudes and for any day of the year. 
  • Multiple pack sizes
  • Recommended for astronomy and earth science at the junior high school through college level
The activity includes data that the students can use to see that the seasons are not caused by the distance between the earth and the sun.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty