Would you like to log in again?
Do-It-Yourself Star Finder Kit
Supplier: SCIENCE INTERACTIVE GROUP
654-0130
470350-110PK 18.79 USD InStock
470350-110
Do-It-Yourself Star Finder Kit
Educational Materials Earth Science Educational Materials Astronomy Learning Activities
Do-It-Yourself Star Finder Kits let your students make their own inexpensive, adjustable star chart that can be set for any time of night on any day of the year using only a pair of scissors and tape. 
  • Packs of 10
  • Each kit include 2 cardboard sheets to make one 8.5" Ø star finder
Students can determine the signs of the zodiac by comparing the constellations visible after sunset and those visible before sunrise. They can even compare the evening sky in all seasons.
The chart shows 41 constellations with lines to indicate the more obvious shapes and patterns. The star images are black on a white sky and study the heifetz planisphere.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty