Would you like to log in again?
Precession of the Equinoxes Kit
Supplier: SCIENCE INTERACTIVE GROUP
654-0160
470350-130EA 46.09 USD InStock
470350-130
Precession of the Equinoxes Kit
Educational Materials Earth Science Educational Materials Astronomy Learning Activities
This unique astronomical device shows which stars are visible at any hour, day or month, thousands of years in the past, the present and thousands of years into the future.
  • Made specifically for the southern hemisphere
  • Created by Dr. Milton D. Heifetz, author of a walk through the heavens: a guide to the stars and constellations and their legends
This unique astronomical device illustrates precession in a way that can otherwise only be duplicated by using a computer and planetarium. It also demonstrates the change in declination over the millennia and which star will be the north start in the future.
The earth orbits around the sun on a plane called the ecliptic. Because of the earth's tilt, twice per year the earth's equatorial plane intersects the ecliptic plane. That time of intersection is called the equinox.
In about 130 B.C., the Greek astronomer Hipparchus noticed a change in the position of the stars. Due to a wobble of the earth as it spins on its axis, the poles of the earth trace a circle in the sky that alters the position of the north and south celestial poles and the celestial equator. As a result, the equinox slowly changes its position along the ecliptic. This is called precession of the equinoxes. 10" Ø 30 to 40 degrees S. latitude.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty