Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Undecanoic acid 99%

Supplier: Ambeed
Warning
Synonyms: Hendecanoic acidUndecylic acid

A1453728-25G
76661-612EA 15.45 USD InStock
76661-612
Undecanoic acid 99%
Undecanoic acid
Caution: for Research Use Only


Formula: C₁₁H₂₂O₂
MW: 186.29 g/mol
MDL Number: MFCD00002730
CAS Number: 112-37-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty